Learn More
Brady™ Nylon Cloth Labels for Portable Label Printers
Autoclavable nylon cloth labels for portable label printers, suitable for low temperatures.
Brand:  Brady™ 152094
 View more versions of this product
Product Code. 17673205
  • 39.50 € / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Nylon cloth labels for Brady™ M210 and M211 portable label printers.
  • Aggressive, permanent adhesive
  • Die-cut labels
  • For low temperatures, such as in freezers or liquid nitrogen
  • Autoclavable
Specifications
Specifications
Label
Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer
Thermal Transfer
50.8 mm
19 mm
Permanent Acrylic
Non-sterile
Cartridge, Label
Component and Equipment Labeling, Electrical Labeling, General Identification, Laboratory Labeling
Black on White
Nylon Cloth
2 in.
0.75 in.
0.165 mm
Autoclavable
Matte
2011/65/EU and 2015/863
73