missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Calibration Buffer Bottles, Oakton™
Use these PH buffers for meters with 0.01 resolution. Oakton™ pH Calibration Buffer Bottles are standardized against NIST-traceable pH references within 0.01 pH at 77°F (25°C).
15.90 € - 29.80 €
Product Code | Brand | pH | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH | Price | Quantity & Availability | |||||
11739679
|
Cole Parmer
00654-01 |
1.68 |
16.10 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10038434
|
Cole Parmer
00654-00 |
4.01 |
29.75 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10303765
|
Cole Parmer
00654-04 |
7.00 |
29.70 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11750079
|
Cole Parmer
00654-08 |
10.00 |
29.80 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11710069
|
Cole Parmer
00654-12 |
12.46 |
15.90 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- pH buffer solutions allow the user to adjust their system to read accurate measurements
- Store buffers away from direct sunlight.
Order Info
Shipping weight: 1-1/4 lbs. (0.6kg).
Certifications
NIST traceable, Certified to NIST traceable standards.
Specifications
500 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- pH 1.68 Buffer
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P202-Do not handle until all safety precautions have been read and understood.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title