missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oakton™ Waterproof Big Display pHTestr™ 30
Versatile, compact design works in a field or laboratory setting.
146.00 €
Specifications
Accuracy (pH) | ±0.01 |
---|---|
Accuracy (Temperature) | ±1°C |
For Use With (Application) | Hold function, auto-off, error messages, waterproof, dustproof and floatable |
Range (pH) | -1 to 15 |
Resolution (pH) | 0.01 |
Includes: Meter Only package includes meter, electrode, four 1.5V batteries, instructions. Meter Kit package includes meter; electrode; two each of pH 4.01, 7.00, 10.00 buffers and rinse solution in single-use pouches, sample jar, batteries, instructions and hard Plastic carrying case
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
13444606
|
Oakton™
35634-30 |
Meter, electrode, four 1.5V batteries, instructions |
146.00 €
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
Description
- Simultaneous pH and temperature display
- Temperature measurement from 0.0° to 50.0°C
- Suitable for most ASTM and EPA test procedures
- Microprocessor-based, with accurate readings to 0.01pH
- Replaceable double-junction electrode (S65290 or NC9830226) has extra reference gel for longer electrode life
- Automatic temperature compensation
- Push-button three-point calibration
- Auto buffer recognition: selectable buffer sets
- Hold function; auto-off; error messages
- Waterproof, dustproof and floatable; certifiable to IP67 standards
Order Info
- Ordering Information
- Meter dimensions: 6-1/2 in.H x 1-1/2 in. dia. (16.5 x 3.8cm). Shipping weight (meter only): 4.5 oz. (125g).
Warranty
One-year warranty.
Specifications
±0.01 | |
Hold function, auto-off, error messages, waterproof, dustproof and floatable | |
0.01 | |
0.1°C | |
USA, NIST | |
1 Year | |
0.1pH | |
32.0°F to 122.0°F | |
16.5 cm | |
3.8 cm |
±1°C | |
-1 to 15 | |
1 to 3 | |
LCD | |
IP-67 | |
±0.1 pH | |
-1 to 15 pH | |
0°C to 50°C | |
1.5 in. |