Learn More
Diba Omnifit™ BenchMark™ Microbore Chromatography Columns
Optimized for aqueous systems and compatible with solvents used in common liquid chromatography applications like protein purification
Brand:  Diba 006BCC-03-10-FF
Encompass_Preferred
 View more versions of this product
Product Code. 15835254
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Includes: Ready-assembled column consisting of glass tube and two fixed endpieces preassembled with 2μm SS frits.
Description
Description
  • Economical, nonadjustable columns for low pressure or gravity feed
  • 3mm diameter and a variety of lengths
  • Compatible with a variety of bulk media
Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, >10% NaOH, >10% HCl, >5% acetic acid, or strong mineral acid.
Specifications
Specifications
3 mm
Chromatography Column
2 μm
100 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Diba Omnifit™ BenchMark™ Microbore Chromatography Columns