Learn More
Thermo Scientific™ Oncomine™ NGS Assay Service
Oncomine NGS Assay Service detects genomic variations associated with select cancer hotspots in iPSC samples.
Brand:  Thermo Scientific™ A46080
Product Code. 18795485
  • 1840.00 € / Each
Explore more special offers
Description
Description
  • Detects genomic variations associated with select cancer hotspots in iPSC samples
  • This service leverages the Ion Torrent Oncomine Comprehensive Assay v3M (A35805) to enable analysis of variants across 161 genes
The Oncomine NGS Assay Service offers:
  • Fast data delivery – turnaround time of four weeks or less
  • Convenient sample prep – just send a frozen cell pellet
  • Detailed results – detection of SNVs, CNVs, gene fusions, and indels
  • Validated data – assay performed in a CLIA-certified clinical laboratory
  • Exceptional support – a dedicated project manager for frequent project communication
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 sample
NGS Assay Service
Cell Line Developers
4 Weeks
Research Use Only
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Oncomine™ NGS Assay Service > 1 Sample