Learn More
Berkshire™ Open-Cell Foam Lab-Tips™ Swabs
General-purpose swab for cleaning small and confined areas
Brand:  Berkshire™ LTO70F.20
 View more versions of this product
Product Code. 17863232
  • 474.00 € / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polyurethane foam over rigid core
  • 100 ppi open-cell foam
  • Thermally bonded head
  • Low particles, fibers, ions and extractables, no contaminating adhesives
  • Good chemical resistance
  • Good sorbency and excellent solvent hold
  • Laundered and packaged in cleanroom
Specifications
Specifications
ISO Class 4
Designed for the highest level of contamination control in critical processing applications, cleaning of sensitive equipment, applying lubricants and other liquids, removing excess materials
71mm
59mm
2 mm
11mm
0.157 in.
2.8 in.
Swab
2.32 in.
Polyurethane Foam
0.43 in.
4 mm
Flexible