Learn More
Product Code | Brand | Length (Metric) | Width (Metric) Tip | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) Tip | Price | Quantity & Availability | ||||||
17833232
|
Berkshire™
LTA03163.10 |
76mm | 5 mm |
641.00 €
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
17897290
|
Berkshire™
LTA03163.10 |
76mm | 5 mm |
723.00 €
Pack of 1000 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
17863232
|
Berkshire™
LTO70F.20 |
71mm | 4 mm |
474.00 €
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
17873232
|
Berkshire™
LTO70P.20 |
71 mm | 3mm |
499.00 €
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
17825510
|
Berkshire™
LTO70P.20 |
71 mm | 3mm |
564.00 €
Pack of 20 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
17883232
|
Berkshire™
LTO70R.20 |
71mm | 3.7mm |
476.00 €
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
12876774
![]() |
Berkshire™
LTO70R.20 |
71mm | 3.7mm |
764.00 €
Pack of 20 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
17853232
|
Berkshire™
LTO1465.10 |
162mm | 3.8mm |
372.00 €
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
12897153
|
Berkshire™
LT000125.10 |
127mm | 13 mm | N/A | N/A | N/A | |||||
Description
- Polyurethane foam over rigid core
- 100 ppi open-cell foam
- Thermally bonded head
- Low particles, fibers, ions and extractables, no contaminating adhesives
- Good chemical resistance
- Good sorbency and excellent solvent hold
- Laundered and packaged in cleanroom
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Berkshire™ Open-Cell Foam Lab-Tips™ Swabs