Learn More
Berkshire™ Open-Cell Foam Lab-Tips™ Swabs
General-purpose swab for cleaning small and confined areas
Brand:  Berkshire™ LTO70R.20
 View more versions of this product
Product Code. 17883232
  • 476.00 € / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polyurethane foam over rigid core
  • 100 ppi open-cell foam
  • Thermally bonded head
  • Low particles, fibers, ions and extractables, no contaminating adhesives
  • Good chemical resistance
  • Good sorbency and excellent solvent hold
  • Laundered and packaged in cleanroom
Specifications
Specifications
ISO Class 4
2.8 in.
Swab
71mm
58mm
3.8mm
12.5mm
1 in.
0.11 in.
Designed for the highest level of contamination control in critical processing applications, cleaning of sensitive equipment, applying lubricants and other liquids, removing excess materials
2.8mm
2.28 in.
Polyurethane Foam
0.49 in.
3.7mm
Rigid