Learn More
Köttermann Open Tall Storage Cabinet
The open-front design of this Open Tall Storage Cabinet provides easy access to stored equipment and utensils.
Brand:  Köttermann 301-00005
 View more versions of this product
Product Code. 17352079
  • 823.00 € / Each
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Tall, slender design expands storage with a small footprint
  • Open front provides easy access to stored items
Specifications
Specifications
White
Tall Storage
None
Storage
None
None
No
No
350 x 1200 x 1920 mm
Epoxy Coated
None
4
69.7 kg
Steel