missing translation for 'onlineSavingsMsg'
Learn More

Köttermann Open Tall Storage Cabinet

The open-front design of this Open Tall Storage Cabinet provides easy access to stored equipment and utensils.

Brand:  Köttermann 301-00018

 View more versions of this product

Product Code. 17372079

  • 1715.00 € / Each
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Tall, slender design expands storage with a small footprint
  • Open front provides easy access to stored items
Specifications

Specifications

White
Tall Storage
None
Storage
4
None
No
No
500 x 1200 x 1920 mm
Epoxy Coated
None
None
106.4 kg
Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.