Learn More
Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles
Control sensitivity, specificity and stability with various acid content available within Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles.
Brand:  Thermo Scientific™ W080CB
 View more versions of this product
Product Code. 11920085
Description
Show More
Specifications
Specifications
Proprietary anionic surfactant
4%
Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications.
Covalent Coupling
Specialty Particles
100 mL
100 mL
Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request.
0.85 μm
Includes Certificate of Analysis
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
White or Colorless
Polystyrene Sulfate
Package Insert Sheet containing Certicate of Analysis
1.05g/cm³
Packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles.
Medium Acid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles > Dia: 0.85μm; 100mL; Medium acid