Learn More
Macherey-Nagel™ Optima™ 1701 High Performance GC Capillary Column
Brand:  Macherey-Nagel™ 726318.50
 View more versions of this product
Product Code. 10420322
  • 1090.00 € / Pack of 50
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
0.5 mm
0.32 mm
G46
300°C to 320°C
50 m
High Performance GC Capillary Column
Optima™ 1701
Gas Chromatography (GC)
0.25 μm
1 Each