Learn More
Macherey-Nagel™ Optima™ 1701 High Performance GC Capillary Column
280.00 € - 1160.00 €
Specifications
Stationary Phase | Optima™ 1701 |
---|---|
USP Type | G46 |
Type | High Performance GC Capillary Column |
For Use With (Application) | Gas Chromatography (GC) |
Quantity | 1 Each |
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||
10506511
|
Macherey-Nagel™
726058.10 |
10 m | 0.25 mm | 0.25 μm |
280.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10328071
|
Macherey-Nagel™
726318.10 |
10 m | 0.32 mm | 0.25 μm |
297.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10022931
|
Macherey-Nagel™
726545.10 |
10 m | 0.53 mm | 1 μm |
348.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10526681
|
Macherey-Nagel™
726058.15 |
15 m | 0.25 mm | 0.25 μm |
375.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10671492
|
Macherey-Nagel™
726318.15 |
15 m | 0.32 mm | 0.25 μm |
384.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10758622
|
Macherey-Nagel™
726735.15 |
15 m | 0.53 mm | 0.5 μm |
420.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10052951
|
Macherey-Nagel™
726545.15 |
15 m | 0.53 mm | 1 μm |
420.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10736702
|
Macherey-Nagel™
726058.25 |
25 m | 0.25 mm | 0.25 μm |
605.00 €
Each |
In Stock
Log in to see stock. |
|||
10419751
|
Macherey-Nagel™
726841.25 |
25 m | 0.2 mm | 0.5 μm |
605.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10002911
|
Macherey-Nagel™
726318.25 |
25 m | 0.32 mm | 0.25 μm |
646.00 €
Each |
In Stock
Log in to see stock. |
|||
10032851
|
Macherey-Nagel™
726824.25 |
25 m | 0.32 mm | 0.35 μm |
646.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10526501
|
Macherey-Nagel™
726320.25 |
25 m | 0.32 mm | 0.5 μm |
646.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10505551
|
Macherey-Nagel™
726929.25 |
25 m | 0.32 mm | 1 μm |
646.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10296751
|
Macherey-Nagel™
726735.25 |
25 m | 0.53 mm | 0.5 μm |
749.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10276941
|
Macherey-Nagel™
726058.30 |
30 m | 0.25 mm | 0.25 μm |
605.00 €
Each |
In Stock
Log in to see stock. |
|||
10012921
|
Macherey-Nagel™
726064.30 |
30 m | 0.25 mm | 0.5 μm |
605.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10114711
|
Macherey-Nagel™
726965.30 |
30 m | 0.25 mm | 1 μm |
605.00 €
Each |
In Stock
Log in to see stock. |
|||
10144561
|
Macherey-Nagel™
726318.30 |
30 m | 0.32 mm | 0.25 μm |
646.00 €
Each |
In Stock
Log in to see stock. |
|||
10338401
|
Macherey-Nagel™
726545.25 |
25 m | 0.53 mm | 1 μm |
749.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10776512
|
Macherey-Nagel™
726824.30 |
30 m | 0.32 mm | 0.35 μm |
646.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10032951
|
Macherey-Nagel™
726320.30 |
30 m | 0.32 mm | 0.5 μm |
646.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10429941
|
Macherey-Nagel™
726929.30 |
30 m | 0.32 mm | 1 μm |
646.00 €
Each |
In Stock
Log in to see stock. |
|||
10681492
|
Macherey-Nagel™
726735.30 |
30 m | 0.53 mm | 0.5 μm |
749.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10348401
|
Macherey-Nagel™
726545.30 |
30 m | 0.53 mm | 1 μm |
749.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10491272
|
Macherey-Nagel™
726058.50 |
50 m | 0.25 mm | 0.25 μm |
1005.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10649771
|
Macherey-Nagel™
726841.50 |
50 m | 0.2 mm | 0.5 μm |
1005.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10420322
|
Macherey-Nagel™
726318.50 |
50 m | 0.32 mm | 0.25 μm |
1090.00 €
Pack of 50 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10489551
|
Macherey-Nagel™
726824.50 |
50 m | 0.32 mm | 0.35 μm |
1090.00 €
Each |
In Stock
Log in to see stock. |
|||
10768042
|
Macherey-Nagel™
726320.50 |
50 m | 0.32 mm | 0.5 μm |
1090.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10795932
|
Macherey-Nagel™
726735.50 |
50 m | 0.53 mm | 0.5 μm |
1160.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10523602
|
Macherey-Nagel™
726929.50 |
50 m | 0.32 mm | 1 μm |
1090.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10797272
|
Macherey-Nagel™
726064.60 |
60 m | 0.25 mm | 0.5 μm |
1050.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10516511
|
Macherey-Nagel™
726058.60 |
60 m | 0.25 mm | 0.25 μm |
1050.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10246941
|
Macherey-Nagel™
726318.60 |
60 m | 0.32 mm | 0.25 μm |
1155.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10042851
|
Macherey-Nagel™
726824.60 |
60 m | 0.32 mm | 0.35 μm |
1155.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10246271
|
Macherey-Nagel™
726320.60 |
60 m | 0.32 mm | 0.5 μm |
1155.00 €
Each |
In Stock
Log in to see stock. |
|||
10595381
|
Macherey-Nagel™
726929.60 |
60 m | 0.32 mm | 1 μm |
1155.00 €
Each |
In Stock
Log in to see stock. |
|||
Specifications
Optima™ 1701 | |
High Performance GC Capillary Column | |
1 Each |
G46 | |
Gas Chromatography (GC) |