Learn More
Macherey-Nagel™ Optima™ 1701 High Performance GC Capillary Column
Brand:  Macherey-Nagel™ 726929.50
 View more versions of this product
Product Code. 10523602
  • 1090.00 € / Each
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.5 mm
0.32 mm
G46
300°C to 320°C
50 m
High Performance GC Capillary Column
Optima™ 1701
Gas Chromatography (GC)
1 μm
1 Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Optima™ 1701 High Performance GC Capillary Column > Length: 50m; I.D.: 0.32mm; Film Thickness: 1um