Learn More
Macherey-Nagel™ Optima™ 1701 High Performance GC Capillary Column
Brand:  Macherey-Nagel™ 726841.50
 View more versions of this product
Product Code. 10649771
  • 1005.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.4 mm
0.2 mm
G46
300°C to 320°C
50 m
High Performance GC Capillary Column
Optima™ 1701
Gas Chromatography (GC)
0.5 μm
1 Each