Learn More
Macherey-Nagel™ Optima™ 1701 High Performance GC Capillary Column
Brand:  Macherey-Nagel™ 726735.15
 View more versions of this product
Product Code. 10758622
  • 420.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.8 mm
0.53 mm
G46
280°C to 300°C
15 m
High Performance GC Capillary Column
Optima™ 1701
Gas Chromatography (GC)
0.5 μm
1 Each