Learn More
Macherey-Nagel™ Optima™ 1701 High Performance GC Capillary Column
Brand:  Macherey-Nagel™ 726824.30
 View more versions of this product
Product Code. 10776512
  • 646.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.5 mm
0.32 mm
G46
300°C to 320°C
30 m
High Performance GC Capillary Column
Optima™ 1701
Gas Chromatography (GC)
0.35 μm
1 Each