missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ OptiPRO™ SFM
GREENER_CHOICE

OptiPRO™ SFM

Brand:  Gibco™ 12309050

31.99 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10569520

  • 33.67 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

OptiPRO™ SFM is a serum-free, animal origin-free culture medium designed for growth of several kidney-derived cell lines including MDBK, MDCK, PK-15 and VERO for virus or recombinant protein production. In addition, OptiPRO™ SFM has successfully grown other cell lines, such as Hela, BHK-21 and COS-7. Gibco™ OptiPRO™ SFM features:

  • Serum-free, animal origin-free, low protein formulation
  • Direct adaptation for many popular cell lines

Serum-free, animal origin-free, low protein formulation

Gibco™ OptiPRO™ SFM is formulated with a very low protein concentration (< 10 μg/mL), allowing for easier purification of your recombinant protein.

Direct adaptation for many popular cell lines

Gibco™ OptiPRO™ SFM has been designed for growth of kidney-derived cell lines such as MDCK, MDBK, VERO and PK-15. In addition, OptiPRO™ SFM has successfully grown other cell lines, such as Hela, BHK-21 and COS-7, allowing you to choose a single medium for multiple cell lines.

cGMP Manufacturing and Quality System

Gibco™ OptiPRO™ SFM is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HeLa, MDCK, BHK-21, PK-15, COS-7, MDBK, VERO
1 X
Serum Free Medium
Phenol Red, Sodium Pyruvate, Sodium Bicarbonate, Thymidine, Hypoxanthine
Gibco™
Room Temperature
Bottle(s)
Storage conditions: 2 – 8°C. Protect from light.
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Animal Origin-free, Low Protein, Serum-free
Liquid
Serum-free
No Glutamine
100 mL
Low
No antibiotics
Suspension Cell Culture
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3064199 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number12309050,12309019
Lot Number2451723 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12309050,12309019
Lot Number2187161 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12309050,12309019
Lot Number2445645 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12309050,12309019
Lot Number2451087 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12309050,12309019
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ OptiPRO™ SFM > Quantity: 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.