Learn More
Gibco™ OptiPRO™ SFM

OptiPRO™ SFM
33.67 € - 133.00 €
Specifications
Cell Line | HeLa, MDCK, BHK-21, PK-15, COS-7, MDBK, VERO |
---|---|
Classification | Animal Origin-free, Low Protein, Serum-free |
Concentration | 1 X |
Form | Liquid |
Product Type | Serum Free Medium |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10569520
![]() |
Gibco™
12309050 |
100 mL |
33.67 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11530426
![]() |
Gibco™
12309019 |
1000 mL |
133.00 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OptiPRO™ SFM is a serum-free, animal origin-free culture medium designed for growth of several kidney-derived cell lines including MDBK, MDCK, PK-15 and VERO for virus or recombinant protein production. In addition, OptiPRO™ SFM has successfully grown other cell lines, such as Hela, BHK-21 and COS-7. Gibco™ OptiPRO™ SFM features:
- Serum-free, animal origin-free, low protein formulation
- Direct adaptation for many popular cell lines
Serum-free, animal origin-free, low protein formulation
Gibco™ OptiPRO™ SFM is formulated with a very low protein concentration (< 10 μg/mL), allowing for easier purification of your recombinant protein.
Direct adaptation for many popular cell lines
Gibco™ OptiPRO™ SFM has been designed for growth of kidney-derived cell lines such as MDCK, MDBK, VERO and PK-15. In addition, OptiPRO™ SFM has successfully grown other cell lines, such as Hela, BHK-21 and COS-7, allowing you to choose a single medium for multiple cell lines.
cGMP Manufacturing and Quality System
Gibco™ OptiPRO™ SFM is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
HeLa, MDCK, BHK-21, PK-15, COS-7, MDBK, VERO | |
1 X | |
Serum Free Medium | |
Phenol Red, Sodium Pyruvate, Sodium Bicarbonate, Thymidine, Hypoxanthine | |
Room Temperature | |
Bottle(s) | |
Storage conditions: 2 – 8°C. Protect from light. Shipping conditions: Ambient Shelf life: 24 months from date of manufacture |
Animal Origin-free, Low Protein, Serum-free | |
Liquid | |
Serum-free | |
Gibco™ | |
Low | |
No antibiotics | |
Suspension Cell Culture |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.