Learn More
Thermo Scientific™ Orion™ Cupric Electrodes
Measure cupric ions in aqueous solutions quickly, simply, accurately and economically with these cupric electrodes.
Brand: Thermo Scientific™ 9629BNWP
867.00 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
Measure cupric ions in aqueous solutions quickly, simply, accurately and economically with the Thermo Scientific™ Orion™ Cupric Electrodes.
No separate reference electrode is needed with this electrode, making it convenient to use with small sample volumes.- Calibration Standards Catalog number: 0.1 M Cu(NO3)2/942906
- Required ISA Catalog number: 940011
Specifications
Waterproof BNC | |
Free-flowing liquid junction assures stable, drift-free potentials | |
Combination Ion Selective Electrode |
With waterproof BNC connector BNC-to-USS adapter is required with this product as a onetime purchase. Part Number: 13-641-890 | |
ISE Meters |