Learn More
Thermo Scientific™ Orion™ Electrode Adapter Cables
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
Brand: Thermo Scientific™ 91CBNC
This item is not returnable.
View return policy
Description
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
- For use with meters that have BNC inputs and electrodes with screw cap connectors
Specifications
Screw Cap | |
Meters with 2 mm Reference Pins Connectors, Meters with BNC Connectors, Meters with British Connectors, Meters with Certain LEMO Connectors, Meters with E DIN Connectors, Meters with Radiometer Connectors, Meters with Stripped Lead Inputs Connectors, Meters with US Standard Connectors | |
BNC |
BNC; Screw Cap; Detachable Cable to BNC Connector | |
Electrode Adapter Cable |
Product Content Correction
Product Title
Thermo Scientific™ Orion™ Electrode Adapter Cables > BNC; Screw Cap; Detachable Cable to BNC Connector
Spot an opportunity for improvement?Share a Content Correction