Learn More
Thermo Scientific™ Orion™ Electrode Adapter Cables
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
Brand:  Thermo Scientific™ 91USRF
 View more versions of this product
Product Code. 11722867
  • 234.17 € / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
  • For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2mm reference pins or just a stripped lead inputs
  • Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Screw Cap
Meters with 2 mm Reference Pins Connectors, Meters with BNC Connectors, Meters with British Connectors, Meters with Certain LEMO Connectors, Meters with E DIN Connectors, Meters with Radiometer Connectors, Meters with Stripped Lead Inputs Connectors, Meters with US Standard Connectors
2 mm Pin Tip
2mm Pin Tip; Screw Cap; Detachable Cable to 2.5 mm Pin Tip Connector
Electrode Adapter Cable