Learn More
Thermo Scientific™ Orion™ Ion Selective Electrode Accessories
Use Thermo Scientific™ Orion™ Ion Selective Electrode Accessories for solid state and plastic membrane ion selective electrodes.
Brand:  Thermo Scientific™ 9300BNWP
 View more versions of this product
Product Code. 10461213
  • 583.91 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Use Thermo Scientific™ Orion™ Ion Selective Electrode Accessories for solid state and plastic membrane ion selective electrodes.
Electrodes handles are used with plastic membrane combination or half-cell electrodes. Polishing strips help restore the performance of solid-state, half-cell, or combination ion selective electrodes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Handle for ISE Half Cell Plastic Membrane Electrodes wth BNC waterproof connector
Ion Selective Electrode
Ion Selective Electrode Accessories
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.