Learn More
Thermo Scientific™ Orion™ Lead Electrodes
Measure lead ions in aqueous solutions quickly, simply, accurately and economically.
Brand:  Thermo Scientific™ 9482BN
 View more versions of this product
Product Code. 10191251
  • 1206.00 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Measure lead ions in aqueous solutions quickly, simply, accurately and economically with Thermo Scientific™ Orion™ Lead Electrodes.
  • Free-flowing Themo Scientific™ Sure-Flow™ junction assures stable, drift-free potentials
  • When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap
  • Lead Half-Cell Electrode used in conjunction with the Orion 900200 Double Junction Reference electrode offers the benefits of the Sure-Flow™ junction design
  • Calibration standards catalog number: 948206 - 0.1 M Pb(CIO4)2; 948207 - 0.1 M Na2SO4
  • Required ISA Catalog numbers: Consult instruction manual
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BNC
drift-free potentials
ISE Meters
With BNC connector
Free-flowing Sure-Flow™ junction assures stable
Lead Ion Selective Electrode, BNC connector