Learn More
Thermo Scientific™ Orion™ Powder Chemistry Packs
Achieve easier and cleaner measurements with pre-measured and ready-to-use powder chemistry packs.
Brand:  Thermo Scientific™ AC4P95
 View more versions of this product
Product Code. 11707737
  • 49.45 € / Each
Explore more special offers
Description
Description
Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.
  • Equivalent to other widely used and recognized chemistries
  • Compatible with Orion™ AQUAfast™ Colorimeters
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AQUAfast phosphate (ortho) reagent powder packs, 100 tests
Drinking Water: ASTM D515-88 (A) / Standard Methods 4500-P E; Wastewater: Standard Methods 4500-P E
100 Phos3 Powder Packs
100 tests
AQUAfast 24 mm round vials (Cat. No. AC2V24)
Phosphate (ortho)
Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter
Powder Pack
0.06 to 2.5 mg/L PO4
Phosphomolybdenum/Ascorbic Acid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Manuals
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Orion™ Powder Chemistry Packs > Phosphorus, LR
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.