Learn More
Thermo Scientific™ Orion™ Versa Star pH / ISE Measurement Module
Add pH, ion concentration, mV, ORP and temperature measurement capabilities to your Versa Star meter using the Thermo Scientific™ Orion™ Versa Star pH / ISE Measurement Module
Brand: Thermo Scientific™ VSTAR-ISE
Description
Add pH, ion concentration, mV, ORP and temperature measurement capabilities to your Thermo Scientific™ Orion™ Versa Star meter using the Thermo Scientific™ Orion™ Versa Star pH / ISE Measurement Module. Each Orion™ Versa Star meter has four channels that can be customized with interchangeable measurement modules. The Versa Star pH/ISE module offers up to six point pH and ISE calibrations and advanced ISE features including incremental techniques (single and double known addition and subtraction, analate addition and subtraction), low range stability and electrode ID with calibration verification.
Orion Versa Star pH/ISE Measurement Module:
- Measure pH, ion concentration, mV, relative mV or ORP with temperature
- Perform up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
- Perform up to a six point ISE (Ion Selective Electrode) calibration with selectable units of ppm, M, mg/L, %, ppb or none
- Incremental techniques include single known addition, double known addition, single known subtraction, double known subtraction, analate addition and analate subtraction
- Advanced ISE features include non-linear selectable auto-blank, low range stability and electrode ID with calibration verification
- Customize measurements to show calibration history and pH electrode condition or simplify to show an extra large pH or ion concentration reading
- Module offers a 3-year replacement warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
±0.002 pH | |
±0.2 mV or ±0.05% of reading whichever is greater | |
±0.1°C | |
1 point (mV mode) | |
1 to 6 | |
Orion VERSA STAR pH/mV/ISE/Temperature Module | |
12 x 3 x 9.5 cm (4.7 x 1.2 x 3.8 in.) | |
VERSA STAR pH/mV/ISE/Temperature Module | |
IP-54 | |
10 per channel with password protection | |
-2.000 to 20.000 | |
0 to 19999 in ppm, M, mg/L, %, ppb or unitless | |
±2000.0mV | |
Up to 4 significant digits | |
0.1, 0.01, 0.001 | |
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting | |
-5°C to 105deg;C | |
5°C to 45°C | |
0.5 lb. | |
0.2 kg |
±0.2 mV or ±0.05 of reading, whichever is greater | |
Timed endpoint, linear point to point, non-linear selectable auto-blank, low concentration range stability, incremental techniques including single known addition and subtraction, double known addition and subtraction, single analyte addition and subtraction | |
Set point alarm, high/low limit alarm, calibration due alarm | |
1 to 6 | |
1 point relative mV (RmV) mode, 1 point ORP mode | |
pH/ISE Measurement Module | |
BNC (pH or ISE), 8-pin Mini-DIN (temperature), reference pin-tip | |
7.000pH (adjustable), 1.000 ISE (adjustable) | |
pH, mV, relative mV (RmV), ORP or ion concentration (ISE) with temperature | |
Orion Versa Star pH/ISE/mV/RmV/ORP/Temperature Measurement Module | |
pH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe (purchase separately) | |
±2000.0mV | |
5 to 85 %, non-condensing | |
0.1mV | |
0.1°C | |
41°F to 113°F | |
1 point temperature offset calibration | |
Manual or automatic with ATC temperature probe | |
3 year module warranty |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.