missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat anti-Mouse IgG2b Secondary Antibody, FITC

Rat Monoclonal Secondary Antibody

Brand:  Invitrogen™ SA135649

Product Code. 11575090

  • 1254.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Mouse Gamma 2b Heavy Chain of Immunoglobulin (determined by immunodot). positively tested on BALB/c and C57BL/6 mouse sera. The specificity of every rat monoclonal antibody anti-mouse IgG subclasses is determined in a range of optimal concentrations. Increasing the first or the second antibody at an unnecessary too high concentration can induce possible cross-reactions with other mouse IgG subclasses. Avidity on IgG2b: 1 * 1010 M-1. No cross-reactivity with chicken, rabbit, goat, sheep, bovine, horse, dog, swine, baboon IgG and human IgG or IgM (ELISA test). Available unconjugated MON5055, FITC MON5055F, Biotin MON5055B, PE MON5055P.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse IgG2b
Monoclonal
1 mg/mL
PBS with 0.1% sodium azide
Immunoglobulins.
1 mL
Secondary
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C. Store in the dark.
Liquid
Immunohistochemistry (Frozen), ELISA
LO-MG2b-2
FITC
Rat
purified
RUO
Mouse
Secondary Antibody
IgG κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Rat anti-Mouse IgG2b Secondary Antibody, FITC > 1 mL; FITC

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.