missing translation for 'onlineSavingsMsg'
Learn More
Osmolality Standard, 50mOsm/kg H2O, Reagecon™ Product Code.: 13315674

Osmolality Standard, 50mOsm/kg H2O, Reagecon™
BIOPHARMA

Product Code. 13315674 Shop All Reagecon Products
12 x 5 mL
Click to view available options
Quantity:
12 x 5 mL
This item is not returnable. View return policy

Product Code. 13315674

Brand: Reagecon REOSM50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Packaging Ampoules
Quantity 12 x 5 mL
Physical Form Liquid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberOSM5024H1 Certificate TypeCertificate of Analysis Date16/03/2025 Catalog NumberRE-OSM-50
Lot NumberOSM5023H1 Certificate TypeCertificate of Analysis Date20/11/2024 Catalog NumberRE-OSM-50
Lot NumberOSM5022E1 Certificate TypeCertificate of Analysis Date19/09/2023 Catalog NumberRE-OSM-50
Lot NumberOSM5021F1 Certificate TypeCertificate of Analysis Date13/06/2022 Catalog NumberRE-OSM-50
Lot NumberOSM5020M1 Certificate TypeCertificate of Analysis Date24/08/2021 Catalog NumberRE-OSM-50
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Osmolality Standard, 50mOsm/kg H2O, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.