Learn More
Fisherbrand™ Oval PTFE Stir Bar
Provides stable stirring action. Fisherbrand; Oval PTFE Stir Bar povides optimal turbulence for round-bottom flasks and containers.
5.06 € - 37.70 €
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
Color | White |
Description | PTFE encapsulated Alnico V magnet core |
Shape | Oval |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||||
11898872
|
Fisherbrand™
001.620 |
20 mm | 10 mm |
17.95 €
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
11517802
|
Fisherbrand™
001.620 |
20 mm | 10 mm | N/A | N/A | N/A | |||||
11858952
|
Fisherbrand™
001.640.13 |
40 mm | 13 mm | N/A | N/A | N/A | |||||
16285831
|
Fisherbrand™
001.650 |
50 mm | 20 mm |
28.95 €
Pack of 5 |
In Stock
Log in to see stock. ![]() |
||||||
11828882
|
Fisherbrand™
001.640 |
40 mm | 20 mm |
25.60 €
Pack of 5 |
In Stock
Log in to see stock. ![]() |
||||||
10603731
|
Fisherbrand™
001.640 |
40 mm | 20 mm | N/A | N/A | N/A | |||||
11338703
|
Fisherbrand™
001.630 |
30 mm | 16 mm | N/A | N/A | N/A | |||||
11808882
|
Fisherbrand™
001.630 |
30 mm | 16 mm |
32.15 €
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
16295831
|
Fisherbrand™
001.664 |
64 mm | 20 mm |
36.90 €
Pack of 5 |
In Stock
Log in to see stock. ![]() |
||||||
11507802
|
Fisherbrand™
001.615 |
15 mm | 6 mm | N/A | N/A | N/A | |||||
11828952
|
Fisherbrand™
001.615 |
15 mm | 6 mm |
14.05 €
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
11868952
|
Fisherbrand™
001.650.17 |
50 mm | 17 mm |
26.90 €
Pack of 5 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
11838952
|
Fisherbrand™
001.625.10 |
25 mm | 10 mm |
20.60 €
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
10445231
|
Fisherbrand™
001.625 |
25 mm | 12 mm | N/A | N/A | N/A | |||||
11844792
|
Fisherbrand™
001.625 |
25 mm | 12 mm |
20.10 €
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
16245831
|
Fisherbrand™
001.635 |
35 mm | 16 mm |
31.85 €
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
10609162
|
Fisherbrand™
001.670 |
70 mm | 20 mm |
16.61 €
Each |
In Stock
Log in to see stock. ![]() |
||||||
11888872
|
Fisherbrand™
001.610 |
10 mm | 5 mm |
12.90 €
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
11597792
|
Fisherbrand™
001.610 |
10 mm | 5 mm | N/A | N/A | N/A | |||||
Description
- Stable, general-purpose magnetic stir bar
- Provides excellent turbulence for round-bottom flasks and containers
- Autoclavable
Specifications
Autoclavable | |
White | |
Oval |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
PTFE encapsulated Alnico V magnet core | |
Round bottom flasks |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Oval PTFE Stir Bar