missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Oval PTFE Turbo Stir Bars with Rare Earth Core
Fisherbrand™ Oval PTFE Turbo Stir Bars with Rare Earth Core use rare earth magnets to provide significantly increased magnetic strength compared to Alnico magnets.
Brand: Fisherbrand™ 001.620.RE
Description
- Almost totally resistant to demagnetisation
- Manufactured from Virgin PTFE materials with FDA and USP Class VI approvals
Specifications
Autoclavable | |
White | |
20 x 10 x 10 mm | |
10 mm | |
10 mm | |
PTFE | |
Oval |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
PTFE encapsulated Samarium-Cobalt magnet core | |
Round Bottom Flasks | |
PTFE | |
20 mm | |
2/Pk. | |
PTFE Stirrer Bars Oval Rare Earth |