missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs.
Marca: Thermo Scientific™ D1-MTD
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with Owl™ D2 Wide-Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.Especificaciones
Micro well; 15 (1X) teeth; 1.5mm thick | |
Owl | |
15 |
Owl™ D2 Wide Gel Electrophoresis System | |
Comb | |
1.5 mm |