missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System
Run 10 to 80 samples on one gel with this wide-gel electrophoresis system.
854.00 € - 1065.00 €
Specifications
Certifications/Compliance | CE marked |
---|---|
Product Line | Owl |
Sample Size | 10 to 80 |
Volume (Metric) Buffer | 600 mL |
Dimensions (L x W x H) | 7.38 x 7.75 x 4 in. (19 x 20 x 10 cm) |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
12404607
|
Thermo Scientific™
D2-BP |
Wide w/Buffer Exchange Ports |
1065.00 €
Each |
In Stock
Log in to see stock. |
|||||
11997475
|
Thermo Scientific™
D2 |
Wide |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Run 10 to 80 samples on one gel with the Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System.
- Wide gel size allows 15 or 30 samples to be loaded per row with a micro well comb.
- Using special combs and a multichannel pipetter, samples may be loaded directly from a 96-well plate, eight or 12 at a time.
- System is excellent for screening PCR samples or routine DNA fragment analysis.
- Small footprint saves space on lab bench.
- Incorporates Thermo Scientific™ EasyCast™ casting design with external caster—gel tray is fitted with gasket, allowing it to fit snugly into caster and providing a leakproof seal without tape.
- Electrode placement allows gel to be run quickly without compromising resolution.
Includes:
- Buffer chamber
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- EasyCast gasketed UVT gel tray
- External multiple gel caster
- Four combs (two 30-well and two 40-well, 1.5mm thick)
Specifications
CE marked | |
10 to 80 | |
7.38 x 7.75 x 4 in. (19 x 20 x 10 cm) | |
19 cm | |
7.75 in. | |
20 cm | |
4 in. | |
150 V |
Owl | |
600 mL | |
7.38 in. | |
14.4 cm | |
6.5 | |
10.2 cm | |
10 cm |