Learn More
Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts
Replace your UVT Gel Tray, gaskets and external caster with Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts, for use with the Owl D4 System.
Brand:  Thermo Scientific™ D4-GK
 View more versions of this product
Product Code. 12444617
  • 18.00 € / Pack of 2
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Replace your UVT Gel Tray, gaskets and external caster with Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts, for use with the Owl D4 System.
  • System design allows you to run one gel tray (51 samples) or stack two gel trays (102 samples) in the buffer chamber while saving valuable bench space.
  • Each of the (2) U.V. Transmissible (UVT) gel trays, 16 x 17 cm (W x L), accommodates up to three combs, allowing the user to run up to three series of samples of equal distances.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Replacement Gaskets (1 pair)
Owl
17 cm
Owl™ D4 Electrophoresis System
Gasket
16 cm