missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts
Replace your UVT Gel Tray, gaskets and external caster with Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts, for use with the Owl D4 System.
18.00 € - 233.00 €
Specifications
For Use With (Equipment) | Owl™ D4 Electrophoresis System |
---|---|
Product Line | Owl |
Length (Metric) | 17 cm |
Width (Metric) | 16 cm |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12434617
|
Thermo Scientific™
D4-CST |
External caster |
233.00 €
Each |
In Stock
Log in to see stock. |
|||||
12444617
|
Thermo Scientific™
D4-GK |
Replacement Gaskets (1 pair) |
18.00 €
Pack of 2 |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
12454617
|
Thermo Scientific™
D4-UVT |
EasyCast™ gasketed UVT gel tray |
137.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
Replace your UVT Gel Tray, gaskets and external caster with Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts, for use with the Owl D4 System.
- System design allows you to run one gel tray (51 samples) or stack two gel trays (102 samples) in the buffer chamber while saving valuable bench space.
- Each of the (2) U.V. Transmissible (UVT) gel trays, 16 x 17 cm (W x L), accommodates up to three combs, allowing the user to run up to three series of samples of equal distances.
Specifications
Owl™ D4 Electrophoresis System | |
Owl | |
17 cm | |
16 cm |