Learn More
Thermo Scientific™ Owl™ Gel-Drying Kits
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures with these gel-drying kits.
Brand:  Thermo Scientific™ GDF-20
 View more versions of this product
Product Code. 11560435
  • 157.00 € / Each
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures. Thermo Scientific™ Owl™ Gel-Drying Kits provide an inexpensive and simple way to produce a permanent record of gels by air drying them between two sheets of cellophane.
  • Available in minigel and large format sizes
  • Convenient method of preserving gels by air drying them between two sheets of cellophane
  • Beveled-edge design of gel-drying frame makes assembly easy
Includes:
Complete kits include two sets of gel-drying frames, 50 sheets of precut cellophane, two solid back plates, and eight side clips; frames and cellphane are also available separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gel-Drying Kit, Large Format
Gel Drying Kit
24 cm
24 x 24 cm
Each
24 cm