Learn More
Thermo Scientific™ Owl™ Gel-Drying Kits
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures with these gel-drying kits.
Brand: Thermo Scientific™ GDF-10
Description
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures. Thermo Scientific™ Owl™ Gel-Drying Kits provide an inexpensive and simple way to produce a permanent record of gels by air drying them between two sheets of cellophane.
- Available in minigel and large format sizes
- Convenient method of preserving gels by air drying them between two sheets of cellophane
- Beveled-edge design of gel-drying frame makes assembly easy
Includes:
Complete kits include two sets of gel-drying frames, 50 sheets of precut cellophane, two solid back plates, and eight side clips; frames and cellphane are also available separately
Specifications
Gel-Drying Kit, Minigel Format | |
Gel Drying Kit | |
14 cm |
14 x 14 cm | |
Each | |
14 cm |