missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems

Provide faster blots with higher efficiency using Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems.

Brand:  Thermo Scientific™ HEP-1

 View more versions of this product

Product Code. 12064927

  • 1215.00 € / Each
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Provide faster blots with higher efficiency using Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems. Transfers can be done in as little as 15 minutes.

  • Fast, efficient transfer of proteins and nucleic acids to membranes
  • Buffer contained within blotting paper is sufficient for efficient transfer of molecules
  • Base contains stainless-steel cathode plate; lid has platinum/titanium anode plate
  • Ideal for proteins less than 150 kDa

Includes:

  • Base and lid
  • Power supply leads
  • Heavy-duty knobs
TRUSTED_SUSTAINABILITY
Specifications

Specifications

For 1 mini gel: Transfer area 20 x 20cm
Owl
9.86 x 9.86 x 2 in. (25 x 25 x 5 cm)
Mini (20 cm x 20 cm)
Electroblotting System
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers