Learn More
Thermo Scientific™ P2-CST Multiple Gradient Gel Caster Comb, 25 teeth
Cast one to five gels simultaneously with the Fisherbrand™ P2-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
14.10 € - 600.00 €
Product Code | Brand | Type | Teeth Count | Thickness (Metric) Teeth | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Teeth Count | Thickness (Metric) Teeth | Price | Quantity & Availability | ||||||
11963055
|
Thermo Scientific™
P2-18G |
Offset Glass | Not Applicable | Not Applicable | N/A | N/A | N/A | |||||
12484677
|
Thermo Scientific™
P2-20FG |
Frosted Blank Glass | Not Applicable | Not Applicable |
42.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
11993055
|
Thermo Scientific™
P2-20G |
Blank Glass | Not Applicable | Not Applicable |
17.80 €
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
12414687
|
Thermo Scientific™
P2-CST |
Caster | Not Applicable | Not Applicable |
600.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
Description
Cast one to five gels simultaneously with the Fisherbrand™ P2-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
- With an external gradient maker, gels can be cast with ease and great reproducibility
- The design and format of this unit will provide years of service in your lab
Specifications
Fisherbrand P2-CST System |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ P2-CST Multiple Gradient Gel Caster Comb, 25 teeth