missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ Sequencing System Spacer Sets and Glass Plates

Use Thermo Scientific™ Owl™ Sequencing System Spacer Sets and Glass Plates with Owl™ Sequencing Gel Casting Systems.

Brand:  Thermo Scientific™ S2S-43G

 View more versions of this product

Product Code. 11997505

  • 27.20 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Use Thermo Scientific™ Owl™ Sequencing System Spacer Sets and Glass Plates with Owl™ Sequencing Gel Casting Systems.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Offset Glass; 43cmL x 35cmW x 0.18 in. thickness
0.32 cm
Offset Glass
35 cm
Owl™ Sequencing System
43 cm
3/16 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Owl™ Sequencing System Spacer Sets and Glass Plates > Offset Glass; 43cmL x 35cmW x 0.18 in. thickness

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.