Learn More
Oxalyl chloride, 2.0M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 369188000
181.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagentSpecifications
Oxalyl chloride | |
79-37-8, 75-09-2 | |
1.3350g/mL | |
C2Cl2O2 | |
MFCD00000704 | |
02, 542 | |
15, 7013 | |
oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2 | |
CTSLXHKWHWQRSH-UHFFFAOYSA-N | |
oxalyl dichloride | |
65578 | |
Liquid |
2.0M solution in dichloromethane | |
Colorless to Yellow | |
AcroSeal™ Glass Bottle | |
ClCOCOCl | |
800 mL | |
01,767; 02,301; 03,216; 04,361; 05,481; 06,424; 07,257; 08,366; 11,379; 14,150; 16,149; 17,241 | |
1.335 | |
Solubility in water: reacts | |
C(=O)(C(=O)Cl)Cl | |
126.93 | |
126.93 |
Safety and Handling
- Oxalyl chloride, 2.0M solution in dichloromethane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H331-Toxic if inhaled.
- H302-Harmful if swallowed.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P402+P404-Store in a dry place. Store in a closed container.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only