Learn More
Oxalyl chloride, 2.0M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals
50.85 € - 181.91 €
Specifications
Chemical Name or Material | Oxalyl chloride |
---|---|
Name Note | 2.0M solution in dichloromethane |
CAS | 79-37-8, 75-09-2 |
Color | Colorless to Yellow |
Density | 1.3350g/mL |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10363372
|
Thermo Scientific Acros
369181000 |
100 mL | AcroSeal™ Glass bottle |
50.85 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10580362
|
Thermo Scientific Acros
369188000 |
800 mL | AcroSeal™ Glass Bottle |
181.91 €
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagentSpecifications
Oxalyl chloride | |
79-37-8, 75-09-2 | |
1.3350g/mL | |
ClCOCOCl | |
02, 542 | |
15, 7013 | |
oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2 | |
CTSLXHKWHWQRSH-UHFFFAOYSA-N | |
oxalyl dichloride | |
65578 | |
Liquid |
2.0M solution in dichloromethane | |
Colorless to Yellow | |
C2Cl2O2 | |
MFCD00000704 | |
01,767; 02,301; 03,216; 04,361; 05,481; 06,424; 07,257; 08,366; 11,379; 14,150; 16,149; 17,241 | |
1.335 | |
Solubility in water: reacts | |
C(=O)(C(=O)Cl)Cl | |
126.93 | |
126.93 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Reacts violently with water.
Contact with water liberates toxic gas.
Corrosive to t
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-200-2
Recommended Storage : Refrigerator +4°C
- Oxalyl chloride, 2.0M solution in dichloromethane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H331-Toxic if inhaled.
- H302-Harmful if swallowed.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only