missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Buffered Peptone Water (Dehydrated)

Pre-enrich food samples prior to the isolation of Salmonella species while allowing the resuscitation of sub-lethally injured cells with this medium.

Brand:  Thermo Scientific™ CM0509B

 View more versions of this product

Product Code. 10036392

  • 110.00 € / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Pre-enrich food samples prior to the isolation of Salmonella species using Thermo Scientific™ Oxoid™ Buffered Peptone Water (Dehydrated). Buffered Peptone Water is a non-selective pre-enrichment medium. It also provides conditions for resuscitation of cells that have been injured by processes of food preservation.

Low numbers and/or sub-lethally injured Salmonella spp. cells may be present in a test food sample. Pre-enrichment in non-selective Buffered Peptone Water allows the cells time to repair and multiply before being introduced into a selective culture, improving the chances of recovering Salmonella species from the sample.

  • Supports high growth and recovery rate: rich in nutrients, the medium produces high resuscitation rates and supports intense growth.
  • Helps recovery of sub-lethally damaged salmonellae: the phosphate buffer system of the medium prevents bacterial damage due to changes in pH of the medium and helps the recovery of sub-lethally damaged salmonellae that may be sensitive to low pH.

It was noted by Edel and Kampelmacher1 that sub-lethal injury to salmonellae may occur in many food processes. In a survey involving isolation of salmonellae from meat that had been artificially contaminated with sub-lethally injured organisms, pre-enrichment in buffered peptone water at 37°C for 18 hours before selection in Brilliant Green Tetrathionate Bile Broth showed superior results compared with a direct selection method.

Sadovski2 reported that, in experiments involving isolation of salmonellae from frozen vegetables, the rapid drop in pH when using lactose broth3 as a pre-enrichment medium was detrimental to the recovery of salmonellae. This was due to the enhanced sensitivity to low pH of freeze-injured salmonellae which may contaminate frozen vegetables. Pre-enrichment with Buffered Peptone Water maintained a high pH over a period of 24 hours of incubation. The vegetable tissue has a low buffering capacity and the medium overcame this problem.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Buffered Peptone Water
Pre-enrichment Medium
For 25.0L medium
Powder
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6248482 Certificate TypeCertificate of Analysis Date04/04/2025 Catalog NumberCM0509B
Lot Number6248440 Certificate TypeCertificate of Analysis Date04/04/2025 Catalog NumberCM0509B
Lot Number6248481 Certificate TypeCertificate of Analysis Date04/04/2025 Catalog NumberCM0509B
Lot Number6224124 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog NumberCM0509B
Lot Number6230077 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog NumberCM0509B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Buffered Peptone Water (Dehydrated) > Buffered Peptone Water; 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use Only