Learn More
Thermo Scientific™ Oxoid™ Minerals Modified Medium (Durhams Tube)
Recommended for the enumeration of coliforms in water and is not intended for use in the diagnosis of disease or other conditions in humans.
Brand:  Thermo Scientific™ EB0542E
Product Code. 12927068
Description
Description
Recommended for the enumeration of coliforms in water and is not intended for use in the diagnosis of disease or other conditions in humans.
2 x strength Oxoid
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4482404 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog NumberEB0542E
Lot Number4482403 Certificate TypeCertificate of Analysis Date09/12/2024 Catalog NumberEB0542E
Lot Number4479279 Certificate TypeCertificate of Analysis Date12/11/2024 Catalog NumberEB0542E
Lot Number4477502 Certificate TypeCertificate of Analysis Date28/10/2024 Catalog NumberEB0542E
Lot Number4471191 Certificate TypeCertificate of Analysis Date04/09/2024 Catalog NumberEB0542E
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Minerals Modified Medium (Durhams Tube) >