missing translation for 'onlineSavingsMsg'
Learn More
p-Dithiane-2,5-diol, 96%, Thermo Scientific Chemicals Product Code.: 11966811

p-Dithiane-2,5-diol, 96%, Thermo Scientific Chemicals

Product Code. 11966811
1 kg, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 11966811

Brand: Thermo Scientific Acros 179930010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40018-26-6
Molecular Formula C4H8O2S2
Molecular Weight (g/mol) 152.23
InChI Key YUIOPHXTILULQC-UHFFFAOYSA-N
Synonym p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anmeShow MoreShow Less
PubChem CID 98330
IUPAC Name 1,4-dithiane-2,5-diol
SMILES C1C(SCC(S1)O)O

Specifications

Melting Point 130.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Plastic bottle
Quantity 1 kg
Solubility Information Solubility in water: 3.9g/L (20°C).
Formula Weight 152.23
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material p-Dithiane-2,5-diol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0425909 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number179932500,179930010
Lot NumberA0425909 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number179932500,179930010
Lot NumberA0402501 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number179932500,179930010
Lot NumberA0327863 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number179932500,179930010
Lot NumberA0381302 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number179932500,179930010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Dithiane-2,5-diol, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.