missing translation for 'onlineSavingsMsg'
Learn More
1,3-Dithiane, 98%, Thermo Scientific Chemicals Product Code.: 11448753

1,3-Dithiane, 98%, Thermo Scientific Chemicals

Product Code. 11448753
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11448753

Brand: Thermo Scientific Alfa Aesar A10505.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,3-Dithiane is used in umpolung reactions such as the Corey-Seebach reaction. It acts as a reagent for the deoxygenation of sulfoxides to their corresponding sulfides. Further, it is used as a protecting group for carbonyl compounds used in organic synthesis. It serves as a useful labeled synthon. In addition to this, it is used in the preparation of 2-ethyl-[1,3]dithiane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dithiane is used in umpolung reactions such as the Corey-Seebach reaction. It acts as a reagent for the deoxygenation of sulfoxides to their corresponding sulfides. Further, it is used as a protecting group for carbonyl compounds used in organic synthesis. It serves as a useful labeled synthon. In addition to this, it is used in the preparation of 2-ethyl-[1,3]dithiane.

Solubility
Soluble in benzene, ether, chloroform and terahydrofuran. Slightly soluble in water.

Notes
Hygroscopic. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-23-7
Molecular Formula C4H8S2
Molecular Weight (g/mol) 120.228
MDL Number MFCD00006654
InChI Key WQADWIOXOXRPLN-UHFFFAOYSA-N
Synonym 1,3-dithiacyclohexane, m-dithiane, dithiane-1,3, 1,3-dithian, dithiane-1,3 french, 1,3 dithiane, ccris 6777, m-dithiane, 8ci, 1,3 dithiocyclohexane, acmc-209knkShow MoreShow Less
PubChem CID 10451
ChEBI CHEBI:47845
IUPAC Name 1,3-dithiane
SMILES C1CSCSC1

Specifications

Melting Point 52°C to 55°C
Boiling Point 207°C to 208°C
Flash Point 90°C (194°F)
Odor Stench
Quantity 5 g
UN Number UN3335
Beilstein 102534
Solubility Information Soluble in benzene,ether,chloroform and terahydrofuran. Slightly soluble in water.
Formula Weight 120.24
Percent Purity 98%
Chemical Name or Material 1,3-Dithiane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM16J035 Certificate TypeCertificate of Analysis Date23/01/2023 Catalog NumberA10505.14,A10505.22,A10505.06
Lot Number10242020 Certificate TypeCertificate of Analysis Date18/07/2022 Catalog NumberA10505.14,A10505.22,A10505.06
Lot NumberI29R023 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10505.14,A10505.22,A10505.06
Lot Number10184440 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10505.14,A10505.22,A10505.06
Lot Number10113609 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10505.14,A10505.22,A10505.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Dithiane, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.