missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ P-Series Ultrasonic Cleaner
Fisherbrand™ P-Series Ultrasonic Water Baths have convenient digital display of set and operating parameters, a cavitation resistant stainless steel tank, ergonomic handles for easy positioning and a durable lid that helps reduce sound and solution evaporation.
Brand: Fisherbrand™ 1013735
1128.38 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
For all applications in analytical, industrial and medical laboratories, including intensive cleaning applications.- Significantly more powerful than conventional ultrasonic units
- Practical and easy to operate with clear display showing set and actual parameters
- Multi-frequency
- 37kHz for standard applications
- 80kHz for prolonged cleaning time, ideal for the treatment of very small and sensitive items
- Units feature normal, pulse, sweep, degas and power control functions
- Side-mounted drain valve and reasr drain duct to remove spent solution
- Powerful pulse mode removes stubborn contaminants
- Stainless steel basket, plastic cover available separately
Order Info
Stainless steel basket, plastic cover available separately
Specifications
V2A Stainless Steel | |
6.9 L | |
230VAC, 970w | |
P 70 H | |
5.6 kg |
230 VAC | |
179 x 568 x 221 mm | |
Professional applications in analytical and medical laboratories and in industrial production processes | |
5°C to 40°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction