Learn More
Bryson Packaging™ Aluminum Foil
Suited for general wrapping purposes and many laboratory applications. Bryson Packaging™ Aluminum Foil is ideal for use in autoclaves. Available in 100mm wide, 18μM guage and 500mm wide, 12μM guage rolls. Each roll has 75m of quality packaging foil.
Brand:  Bryson Packaging™ AKL-310-040C
 View more versions of this product
Product Code. 12309558
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Suitable for general wrapping purposes and many laboratory applications, including:
    • sealing bottles
    • culture tubes
    • stopping leaks
    • reducing evaporation or heat losses
    • covering corks
    • making weighing boats or dishes
  • 75m rolls available in a variety of widths and gaugess
  • Ideal for use in autoclaves
  • Supplied in a cutter box
Specifications
Specifications
Autoclavable
Metallic Silver
100mm
18 μm
246.1 ft.
Aluminum Foil in Cutter Box
Aluminum