Learn More
Bryson Packaging™ Aluminum Foil
Suited for general wrapping purposes and many laboratory applications. Bryson Packaging™ Aluminum Foil is ideal for use in autoclaves. Available in 100mm wide, 18μM guage and 500mm wide, 12μM guage rolls. Each roll has 75m of quality packaging foil.
Brand:  Bryson Packaging™ AKL-300-040J
 View more versions of this product
Product Code. 12379548
  • 267.43 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Suitable for general wrapping purposes and many laboratory applications, including:
    • sealing bottles
    • culture tubes
    • stopping leaks
    • reducing evaporation or heat losses
    • covering corks
    • making weighing boats or dishes
  • 75m rolls available in a variety of widths and gaugess
  • Ideal for use in autoclaves
  • Supplied in a cutter box
Specifications
Specifications
Autoclavable
Metallic Silver
500mm
12 μm
246.1 ft.
Aluminum Foil in Cutter Box
Aluminum
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bryson Packaging™ Aluminum Foil > Dimensions (L x W): 75m x 500mm; 12μm thick