Learn More
Palladium(II) iodide, 99.9% (metals basis), Pd 28% min, Thermo Scientific Chemicals
184.00 € - 913.00 €
Chemical Identifiers
CAS | 7790-38-7 |
---|---|
Molecular Formula | I2Pd |
Molecular Weight (g/mol) | 360.23 |
MDL Number | MFCD00011171 |
InChI Key | HNNUTDROYPGBMR-UHFFFAOYSA-L |
IUPAC Name | palladium(2+) diiodide |
SMILES | [Pd++].[I-].[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11311968
|
Thermo Scientific Alfa Aesar
011881.04 |
2 g |
184.00 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321968
|
Thermo Scientific Alfa Aesar
011881.09 |
10 g |
913.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Palladium(II) iodide is used in gravimetric analysis. It is involved in the synthesis of symmetrical and unsymmetrical ureas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7790-38-7 | |
360.23 | |
HNNUTDROYPGBMR-UHFFFAOYSA-L | |
[Pd++].[I-].[I-] |
I2Pd | |
MFCD00011171 | |
palladium(2+) diiodide |
Specifications
350°C (decomposition) | |
Black | |
I2Pd | |
Soluble in ammonia,methyl acetate and potassium iodide. Slightly soluble in hydriodic acid and concentrated nitric acid. Insoluble in water,ethanol,diluted nitric acid,hydrochloric acid and diethyl ether. | |
[Pd++].[I-].[I-] | |
360.23 | |
99.9% | |
6.003 g/mL | |
Palladium(II) iodide |
7790-38-7 | |
Powder | |
MFCD00011171 | |
HNNUTDROYPGBMR-UHFFFAOYSA-L | |
palladium(2+) diiodide | |
360.23 | |
Pd 28% min. | |
(metals basis) |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P261-P264b-P280-P285-P302+P352-P304+P341-P305+P351+P338-P332+P313-P342+P311-P362-P501c
H315-H319-H334
missing translation for 'einecsNumber' : 232-203-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only