Learn More
Palladium(II) nitrate hydrate, 99.8% (metals basis), Pd 39% min, Thermo Scientific Chemicals
306.53 € - 5719.46 €
Chemical Identifiers
CAS | 207596-32-5 |
---|---|
Molecular Formula | N2O6Pd |
Molecular Weight (g/mol) | 230.43 |
MDL Number | MFCD00011169 |
InChI Key | GPNDARIEYHPYAY-UHFFFAOYSA-N |
Synonym | palladium nitrate, palladium ii nitrate, palladous nitrate, palladium dinitrate, unii-5g27lbz05u, nitric acid, palladium 2+ salt, hydrogen tetranitropalladate ii, palladium 2+ dinitrate, palladiumdinitrate |
PubChem CID | 24932 |
IUPAC Name | palladium(2+);dinitrate |
SMILES | [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11321668
|
Thermo Scientific Alfa Aesar
011035.04 |
2 g |
306.53 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311668
|
Thermo Scientific Alfa Aesar
011035.09 |
10 g |
1448.21 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301668
|
Thermo Scientific Alfa Aesar
011035.18 |
50 g |
5719.46 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Palladium(II) nitrate hydrate is used as a precursor to supported palladium catalysts. It is also used as a catalyst in the conversion of alkene nitration to glycol dinitrates. It is involved in the separation of chlorine and iodine and used as biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
207596-32-5 | |
230.43 | |
GPNDARIEYHPYAY-UHFFFAOYSA-N | |
24932 | |
[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
N2O6Pd | |
MFCD00011169 | |
palladium nitrate, palladium ii nitrate, palladous nitrate, palladium dinitrate, unii-5g27lbz05u, nitric acid, palladium 2+ salt, hydrogen tetranitropalladate ii, palladium 2+ dinitrate, palladiumdinitrate | |
palladium(2+);dinitrate |
Specifications
(decomposition) | |
Brown to Red | |
N2O6Pd | |
MFCD00011169 | |
14,6992 | |
Soluble in dilute nitric acid. | |
[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
230.43 | |
230.43 (Anhydrous) | |
Hygroscopic | |
Palladium(II) nitrate hydrate |
207596-32-5 | |
Crystalline | |
Pd(NO3)2·xH2O | |
UN1477 | |
palladium nitrate, palladium ii nitrate, palladous nitrate, palladium dinitrate, unii-5g27lbz05u, nitric acid, palladium 2+ salt, hydrogen tetranitropalladate ii, palladium 2+ dinitrate, palladiumdinitrate | |
GPNDARIEYHPYAY-UHFFFAOYSA-N | |
palladium(2+);dinitrate | |
24932 | |
99.8% | |
(metals basis), Pd 39% min |
Safety and Handling
GHS H Statement
H272-H315-H319-H317;
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P210-P220-P221-P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
H272-H290-H302-H314-H335
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II
missing translation for 'einecsNumber' : 233-265-8
missing translation for 'rtecsNumber' : QV0560000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.